Learn More
DWK Life Sciences Kimble™ O-Ring Glass Connector Cap
Catalog No. 12141423
Change view
Click to view available options
For Use With (Equipment):
216040 HI-VAC™ Manifold
Catalog No. For Use With (Equipment)
12141423 216040 HI-VAC™ Manifold
1 options
Catalog No. 12141423 Supplier DWK Life Sciences Supplier No. 8518100015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Size 15 Cap and O-ring to seal flasks or 216040 HI-VAC™ manifold.
  • Supplied with one size 116 FKM o-ring
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
For Use With (Equipment) 216040 HI-VAC™ Manifold
Material Glass
Includes FKM O-ring
Type O-ring Connector Cap