Learn More
Cytiva O-ring
Catalog No. p-8004336
Change view
Click to view available options
Size:
37.47 x 5.34mm
Catalog No. Size
501055414 37.47 x 5.34mm
1 options
Catalog No. 501055414 Supplier Cytiva Supplier No. 19068501
Only null left
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
Specifications
For Use With (Equipment) For use with Series XK and K 9
Quantity 1Each
Product Type O-Ring
Material Nitrile Rubber
Size 37.47 x 5.34mm