Learn More
Cytiva O-ring
Catalog No. 501055414
Change view
Click to view available options
Size:
37.47 x 5.34mm
Catalog No. Size
501055414 37.47 x 5.34mm
1 options
Catalog No. 501055414 Supplier Cytiva Supplier No. 19068501
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
Specifications
For Use With (Equipment) For use with Series XK and K 9
Quantity 1Each
Product Type O-Ring
Material Nitrile Rubber
Size 37.47 x 5.34mm