Learn More
Ritmed™ O.R. Towels Xray Detectable Prewashed Towels, Sterile
Catalog No. p-200135822
Change view
Click to view available options
Color:
Blue
Green
White
Includes:
2 Towels
4 Towels
6 Towels
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
21300028 Blue 2 Towels
21300029 Green 2 Towels
21300031 White 2 Towels
21300033 Blue 4 Towels
21300036 White 4 Towels
21300039 Green 6 Towels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21300028 Supplier Ritmed™ Supplier No. 5002BX
Only null left
Designed for surgical settings with cotton muslin operation room towels used to absorb biological fluids
  • Ritmed™ O.R. Towels are specifically designed for use in surgical settings with 17 x 25 in. with 100% cotton and latex free
  • A key safety feature is their X-ray detectable property, which helps prevent items from being left inside a patient's body after surgery
  • The towels are prewashed, making them ready for immediate use and free of any processing debris
  • They are sterile, ensuring a clean and safe product for critical medical procedures
  • This product is an essential tool for surgical staff to maintain a high standard of patient safety
  • The combination of sterility and X-ray detectability highlights the product's focus on quality and risk mitigation
Specifications
Product Type Towel
Color Blue
Material Cotton
Sterility Sterile
Autoclavable Not Autoclavable
Includes 2 Towels
Dimensions (L x W) 17 x 25 in.
Disposable Single-use
Latex Free Yes