missing translation for 'onlineSavingsMsg'
Learn More

o-Phosphoric Acid, 85% w/w, MilliporeSigma™

Catalog No. p-4382309
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX10006 500 mL
MPX10003 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX10006 Supplier Technidata Supplier No. PX10006
Only null left

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Glass Bottle
Assay Percent Range 85%
Quantity 500 mL
Physical Form Liquid
Linear Formula H3PO6
Formula Weight 98
Grade Reagent
Chemical Name or Material o-Phosphoric Acid
Recommended Storage Room Temperature