missing translation for 'onlineSavingsMsg'
Learn More

o-Phosphoric Acid, 85% w/w, ACS, GR, MilliporeSigma™

Catalog No. MPX09956
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX09956 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX09956 Supplier Technidata Supplier No. PX09956

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

pH 0.5
Color Colorless
Melting Point 21°C
Boiling Point 158°C
Density 1.71g/cm3 at 20°C
Packaging Glass Bottle
Vapor Pressure 2 hPa at 20°C
Quantity 500 mL
Physical Form Liquid
Solubility Information Soluble