missing translation for 'onlineSavingsMsg'
Learn More

o-Phosphoric Acid, 85.0% min., ACS, MilliporeSigma™

Catalog No. MPX0995P1
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX0995P1 2.5 L Poly Coated Glass Bottle
MPX09953 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX0995P1 Supplier Technidata Supplier No. PX0995P1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Poly Coated Glass Bottle
Assay Percent Range 85.0% Min.
Quantity 2.5 L
Physical Form Liquid
Linear Formula H3PO4
Formula Weight 98
Grade ACS/NF/USP
Chemical Name or Material o-Phosphoric Acid, For ACS analysis
Recommended Storage Room Temperature