missing translation for 'onlineSavingsMsg'
Learn More

o-Nitroacetophenone, 95%, Thermo Scientific™

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128320025
SDS View Documents
Thermo Scientific Chemicals
128320025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC128320200
SDS View Documents
Thermo Scientific Chemicals
128320200
20g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

577-59-3
165.15
SUGXZLKUDLDTKX-UHFFFAOYSA-N
11346
CC(=O)C1=CC=CC=C1[N+]([O-])=O
C8H7NO3
MFCD00007144
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
1-(2-nitrophenyl)ethanone

Specifications

577-59-3
100.0
>112°C
Glass bottle
1.5500 to 1.5520
MFCD00007144
07, 288
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
CC(=O)C1=CC=CC=C1[N+]([O-])=O
165.15
165.15
o-Nitroacetophenone, 95%
94.0
159.0°C (16.0 mmHg)
94% min. (GC)
C8H7NO3
O2NC6H4COCH3
2.5g
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
SUGXZLKUDLDTKX-UHFFFAOYSA-N
1-(2-nitrophenyl)ethanone
11346
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-414-8

RTECSNumber : AM9600000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only