missing translation for 'onlineSavingsMsg'
Learn More

O-Methylhydroxylamine Hydrochloride 97.0+%, TCI America™

Supplier:  TCI America M0343100G

 View more versions of this product

Catalog No. M0343100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

O-Methylhydroxylamine Hydrochloride
149°C
CH6ClNO
100 g
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
CON.Cl
83.515
83.52
Crystalline Powder
593-56-6
White
MFCD00012951
3261
XNXVOSBNFZWHBV-UHFFFAOYSA-N
O-methylhydroxylamine;hydrochloride
521874
≥97.0% (N,T)

Chemical Identifiers

593-56-6
83.515
XNXVOSBNFZWHBV-UHFFFAOYSA-N
521874
CON.Cl
CH6ClNO
MFCD00012951
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
O-methylhydroxylamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NC3980000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions