missing translation for 'onlineSavingsMsg'
Learn More

O-Methylhydroxylamine Hydrochloride 97.0+%, TCI America™

Catalog No. M034325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M034325G 25 g
M0343100G 100 g
M0343500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M034325G Supplier TCI America Supplier No. M034325G

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 149°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 83.52
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Methylhydroxylamine Hydrochloride
RTECSNumber NC3980000
TSCA Yes