missing translation for 'onlineSavingsMsg'
Learn More

O-Methylhydroxylamine Hydrochloride (ca. 40% in Water, ca. 5.4mol/L), TCI America™

Catalog No. M088625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M088625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M088625ML Supplier TCI America Supplier No. M088625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Quantity 25 mL
UN Number 3265
Formula Weight 83.52
Physical Form Liquid
Chemical Name or Material O-Methylhydroxylamine Hydrochloride (ca. 40% in Water, ca. 5.4mol/L)
RTECSNumber NC3980000
TSCA Yes