missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Methyl-L-tyrosine, 98%

Catalog No. AC169752500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169752500 250 mg Glass Bottle
AC169750010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169752500 Supplier Thermo Scientific Chemicals Supplier No. 169752500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6230-11-1 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6230-11-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002604
InChI Key GEYBMYRBIABFTA-IMWMWJONNA-N
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
PubChem CID 2723935
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2CH(NH2)CO2H
Beilstein 14, I, 668
Specific Rotation -7° to -9° (20°C, 589nm) (c=2, 1N HCl)
Specific Rotation Condition −8° (20°C c=2,1N HCl)
Quantity 250 mg
Formula Weight 195.22
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less

RUO – Research Use Only