missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Methyl-L-tyrosine, 98%
CAS: 6230-11-1 | C5H8O5 | 148.11 g/mol
$63.71 - $212.40
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6309603
|
Thermo Scientific Chemicals
H6309603 |
1 g |
Each for $63.71
|
|
|||||
|
AAH6309606
|
Thermo Scientific Chemicals
H6309606 |
5 g |
Each for $212.40
|
|
|||||
Description
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-Methyl-L-tyrosine, is used as a fine chemical intermediate.
Solubility
Very soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 6230-11-1 | |
| MFCD00002604 | |
| CHEBI:29003 |
Specifications
| 259°C to 261°C (decomposition) | |
| C5H8O5 | |
| 2212726 | |
| Very soluble in water. | |
| CC(CC(=O)O)(C(=O)O)O | |
| 1 g | |
| 441696 | |
| 195.22 | |
| Solid |
| 6230-11-1 | |
| MFCD00002604 | |
| s-2-hydroxy-2-methylsuccinic acid, l-citramalic acid, s-+-citramalic acid, s-citramalic acid, s-2-methylmalic acid, unii-b3fc6pa80i, b3fc6pa80i, 2s-2-hydroxy-2-methylbutanedioic acid, l-alpha-hydroxypyrotartaric acid, 2s-2-hydroxy-2-methylsuccinic acid | |
| XFTRTWQBIOMVPK-YFKPBYRVSA-N | |
| (2S)-2-hydroxy-2-methylbutanedioic acid | |
| 148.11 | |
| CHEBI:29003 | |
| 98% | |
| O-Methyl-L-tyrosine |
Safety and Handling
EINECSNumber : 228-333-9
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only