missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl-L-tyrosine, 98%

Catalog No. AAH6309603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309603 1 g
AAH6309606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6309603 Supplier Thermo Scientific Chemicals Supplier No. H6309603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6230-11-1 | C5H8O5 | 148.11 g/mol

O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

Solubility
Very soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 6230-11-1
MDL Number MFCD00002604
ChEBI CHEBI:29003

Specifications

Melting Point 259°C to 261°C (decomposition)
Molecular Formula C5H8O5
Beilstein 2212726
Synonym s-2-hydroxy-2-methylsuccinic acid, l-citramalic acid, s-+-citramalic acid, s-citramalic acid, s-2-methylmalic acid, unii-b3fc6pa80i, b3fc6pa80i, 2s-2-hydroxy-2-methylbutanedioic acid, l-alpha-hydroxypyrotartaric acid, 2s-2-hydroxy-2-methylsuccinic acid
Solubility Information Very soluble in water.
InChI Key XFTRTWQBIOMVPK-YFKPBYRVSA-N
SMILES CC(CC(=O)O)(C(=O)O)O
IUPAC Name (2S)-2-hydroxy-2-methylbutanedioic acid
Quantity 1 g
Molecular Weight (g/mol) 148.11
PubChem CID 441696
Formula Weight 195.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less
EINECSNumber 228-333-9
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only