missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O-Methyl-L-tyrosine, 98%

Catalog No. AC169750050
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC169750050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169750050 Supplier Thermo Scientific™ Supplier No. 169750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 6230-11-1
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C10H13NO3
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CH(NH2)CO2H
MDL Number MFCD00002604
Beilstein 14, I, 668
Specific Rotation -7° to -9° (20°C, 589nm) (c=2, 1N HCl)
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
InChI Key GEYBMYRBIABFTA-UHFFFAOYNA-N
SMILES COC1=CC=C(C=C1)CC(C(=O)O)N
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
Specific Rotation Condition −8° (20°C c=2,1N HCl)
Molecular Weight (g/mol) 195.22
Quantity 5g
PubChem CID 2723935
Formula Weight 195.22
Percent Purity ≥97.5%
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less

RUO – Research Use Only