missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Isobutylhydroxylamine Hydrochloride 98.0+%, TCI America™
$231.14 - $687.84
Chemical Identifiers
| CAS | 6084-58-8 |
|---|---|
| Molecular Formula | C4H12ClNO |
| Molecular Weight (g/mol) | 125.596 |
| MDL Number | MFCD00060208 |
| InChI Key | WOXGREIMSJFDPG-UHFFFAOYSA-N |
| Synonym | Isobutyloxyamine Hydrochloride |
| PubChem CID | 12696238 |
| IUPAC Name | O-(2-methylpropyl)hydroxylamine;hydrochloride |
| SMILES | CC(C)CON.Cl |
Chemical Identifiers
| 6084-58-8 | |
| 125.596 | |
| WOXGREIMSJFDPG-UHFFFAOYSA-N | |
| 12696238 | |
| CC(C)CON.Cl |
| C4H12ClNO | |
| MFCD00060208 | |
| Isobutyloxyamine Hydrochloride | |
| O-(2-methylpropyl)hydroxylamine;hydrochloride |
Specifications
| 6084-58-8 | |
| White-Yellow | |
| MFCD00060208 | |
| Isobutyloxyamine Hydrochloride | |
| CC(C)CON.Cl | |
| 125.596 | |
| 125.60 | |
| Crystalline Powder |
| 129°C | |
| C4H12ClNO | |
| 1 g | |
| WOXGREIMSJFDPG-UHFFFAOYSA-N | |
| O-(2-methylpropyl)hydroxylamine;hydrochloride | |
| 12696238 | |
| ≥98.0% (N) | |
| O-Isobutylhydroxylamine Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator