missing translation for 'onlineSavingsMsg'
Learn More

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. E09165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09165G 5 g
E09161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09165G Supplier TCI America Supplier No. E09165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136849-72-4
Molecular Formula C10H17BF4N4O3
Molecular Weight (g/mol) 328.07
MDL Number MFCD00192127 MFCD00192127
InChI Key FPQVGDGSRVMNMR-UHFFFAOYSA-N
Synonym totu, o-cyano ethoxycarbonyl methylenamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino n,n,n',n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n',n'-tetra methyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium bf4, e-2-1-cyano-2-ethoxy-2-oxoethylideneamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n'n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyl uronium tetrafluoroborate, o-ethoxycarboxyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium tetrafluoroborate
PubChem CID 9601231
IUPAC Name ethyl cyano({[(dimethylamino)(dimethyliminiumyl)methoxy]imino})formate; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N

Specifications

Melting Point 144°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 328.07
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator