missing translation for 'onlineSavingsMsg'
Learn More

O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 98+%

Catalog No. p-7035960
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5419403 1 g
AAH5419406 5 g
AAH5419414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5419403 Supplier Thermo Scientific Chemicals Supplier No. H5419403
Only null left

CAS: 125700-73-4 | C14H20BF4N3O3 | 365.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 125700-73-4
Molecular Formula C14H20BF4N3O3
Molecular Weight (g/mol) 365.14
MDL Number MFCD00167753
InChI Key DMTWWFCCBWWXSA-UHFFFAOYNA-N
Synonym 2-endo-5-norbornene-2,3-dicarboxymido-1,1,3,3-tetramethyluronium tetrafluoroborate, dimethylamino 1r,2s,6r,7s-3,5-dioxo-4-azatricyclo 5.2.1.0_,? dec-8-en-4-yl oxy methylidene dimethylazanium tetrafluoroborate
PubChem CID 91634054
IUPAC Name [(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)C2C3CC(C=C3)C2C1=O)=[N+](C)C

Specifications

Melting Point 216°C to 219°C
Quantity 1 g
Sensitivity Light and moisture sensitive
Formula Weight 365.13
Percent Purity ≥98%
Chemical Name or Material O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only