missing translation for 'onlineSavingsMsg'
Learn More

O-(Diphenylphosphinyl)hydroxylamine, 96%

Catalog No. p-7084043
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461850010 1 g
AC461850050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461850010 Supplier Thermo Scientific Chemicals Supplier No. 461850010

CAS: 72804-96-7 | C12H12NO2P | 233.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72804-96-7
Molecular Formula C12H12NO2P
Molecular Weight (g/mol) 233.21
InChI Key SJECIYLGISUNRO-UHFFFAOYSA-N
Synonym o-diphenylphosphinylhydroxylamine, aminooxy diphenylphosphine oxide, o-diphenylphosphinyl hydroxylamine, o-diphenylphosphoryl hydroxylamine, amino diphenylphosphinate, hydroxylamine, o-diphenylphosphinyl, unii-2o9pf20t75, aminooxy diphenyl phosphine oxide, o-diphenylphosphinyl hydroxylamine mi
PubChem CID 10955453
IUPAC Name O-diphenylphosphorylhydroxylamine
SMILES NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 104.0°C
Assay Percent Range 88% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.21
Percent Purity 96%
Chemical Name or Material O-(Diphenylphosphinyl)hydroxylamine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only