missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein

Catalog No. AAA1289918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1289918 50 g
AAA1289909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1289918 Supplier Thermo Scientific Chemicals Supplier No. A1289918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 596-27-0 | C22H18O4 | 346.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 596-27-0
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.38
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
Synonym o-cresolphthalein, cresolphthalein, 3,3'-dimethylphenolphthalein, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-3-methylphenyl, 3',3-dimethylphenolphthalein, o-cresolphalein, 3,3-bis 4-hydroxy-3-methylphenyl isobenzofuran-1 3h-one, unii-xou2d9049g, phenolphthalein, 3',3-dimethyl, 3,3-bis 4-hydroxy-m-tolyl phthalide
PubChem CID 68995
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 223°C to 225°C
Quantity 50 g
Beilstein 310554
Merck Index 14,2576
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-2-benzofuran-1-one
Formula Weight 346.39
Chemical Name or Material o-Cresolphthalein
EINECSNumber 209-881-8
RTECSNumber SM8390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only