missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein, TCI America™

Catalog No. C040425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C040425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C040425G Supplier TCI America Supplier No. C040425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 596-27-0
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.38
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
Synonym o-cresolphthalein, cresolphthalein, 3,3'-dimethylphenolphthalein, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-3-methylphenyl, 3',3-dimethylphenolphthalein, o-cresolphalein, 3,3-bis 4-hydroxy-3-methylphenyl isobenzofuran-1 3h-one, unii-xou2d9049g, phenolphthalein, 3',3-dimethyl, 3,3-bis 4-hydroxy-m-tolyl phthalide
PubChem CID 68995
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 223°C
Quantity 25 g
Formula Weight 346.38
Physical Form Crystalline Powder
Chemical Name or Material o-Cresolphthalein
RTECSNumber SM8390000
TSCA Yes