missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich O-(Carboxymethyl)hydroxylamine hemihydrochloride

Catalog No. C1340810G
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1340810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1340810G Supplier Sigma Aldrich Supplier No. C1340810G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-(Carboxymethyl)hydroxylamine hemihydrochloride

Specifications

CAS 2921-14-4
Melting Point 156°C (lit.)
Molecular Formula C2H5NO3 -+ 0.5 HCl
Linear Formula NH2OCH2COOH -+ 0.5 HCl
MDL Number MFCD00012955
Quantity 10 g
Synonym (Aminooxy)acetic acid hemihydrochloride; (Carboxymethoxy)amine hemihydrochloride; Hydroxylamine-O-acetic acid hemihydrochloride
Molecular Weight (g/mol) 109.3
Percent Purity 98%