missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Butylhydroxylamine Hydrochloride 98.0+%, TCI America™
$108.11 - $326.21
Chemical Identifiers
| CAS | 4490-82-8 |
|---|---|
| Molecular Formula | C4H12ClNO |
| Molecular Weight (g/mol) | 125.596 |
| MDL Number | MFCD00487631 |
| InChI Key | HUYRNQWVAPCTQZ-UHFFFAOYSA-N |
| PubChem CID | 12283956 |
| IUPAC Name | O-butylhydroxylamine;hydrochloride |
| SMILES | CCCCON.Cl |
Chemical Identifiers
| 4490-82-8 | |
| 125.596 | |
| HUYRNQWVAPCTQZ-UHFFFAOYSA-N | |
| O-butylhydroxylamine;hydrochloride |
| C4H12ClNO | |
| MFCD00487631 | |
| 12283956 | |
| CCCCON.Cl |
Specifications
| 4490-82-8 | |
| White-Yellow | |
| MFCD00487631 | |
| HUYRNQWVAPCTQZ-UHFFFAOYSA-N | |
| O-butylhydroxylamine;hydrochloride | |
| 12283956 | |
| ≥98.0% (N,T) | |
| O-Butylhydroxylamine Hydrochloride |
| 159°C | |
| C4H12ClNO | |
| 1 g | |
| CCCCON.Cl | |
| 125.596 | |
| 125.60 | |
| Crystalline Powder |
Safety and Handling
TSCA : No