missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine, 97%

Catalog No. p-3200935
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC382050050 5 g Glass bottle
AC382050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC382050050 Supplier Thermo Scientific Chemicals Supplier No. 382050050
Only null left

CAS: 622-33-3 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-33-3
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00221709
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
PubChem CID 102313
IUPAC Name hydrogen O-benzylhydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.1900g/mL
Boiling Point 110°C to 112°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Linear Formula C6H5CH2ONH2
Quantity 5 g
Beilstein 06, IV, 2562
Specific Gravity 1.19
Solubility Information Solubility in water: soluble
Formula Weight 123.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only