missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine, 96%

Catalog No. p-7046247
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1360106 5 g
AAL1360114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1360106 Supplier Thermo Scientific Chemicals Supplier No. L1360106
Only null left

CAS: 622-33-3 | C7H9NO | 123.155 g/mol

O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.

Solubility
Soluble in dimethylsulfoxide and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 622-33-3
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00221709
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
PubChem CID 102313
IUPAC Name hydrogen O-benzylhydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=CC=CC=C1

Specifications

Density 1.058
Boiling Point 118°C to 119°C (30 mmHg)
Refractive Index 1.541
Linear Formula C6H5CH2ONH2
Quantity 5 g
UN Number UN2735
Beilstein 1906691
Solubility Information Soluble in dimethylsulfoxide and methanol.
Formula Weight 123.15
Percent Purity 96%
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only