missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-serine 99.0+%, TCI America™

Catalog No. B08611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08611G 1 g
B08615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08611G Supplier TCI America Supplier No. B08611G
Only null left

Chemical Identifiers

CAS 4726-96-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00065937
InChI Key IDGQXGPQOGUGIX-VIFPVBQESA-N
Synonym o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
PubChem CID 78457
IUPAC Name (2S)-2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-L-serine