missing translation for 'onlineSavingsMsg'
Learn More

O-Benzoyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B32395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32395G 5 g
B32391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32395G Supplier TCI America Supplier No. B32395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27130-46-7
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.623
InChI Key FTCDQNQIGUCFSA-UHFFFAOYSA-N
PubChem CID 11615307
IUPAC Name methylamino benzoate;hydrochloride
SMILES CNOC(=O)C1=CC=CC=C1.Cl

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 187.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzoyl-N-methylhydroxylamine Hydrochloride