missing translation for 'onlineSavingsMsg'
Learn More

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B48051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48051G 1 g
B48055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48051G Supplier TCI America Supplier No. B48051G
Only null left

Chemical Identifiers

CAS 105379-24-6
Molecular Formula C15H20F6N5OP
Molecular Weight (g/mol) 431.323
MDL Number MFCD00191770
InChI Key XKTRAGMCMJYRRN-UHFFFAOYSA-N
Synonym hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
PubChem CID 13685355
IUPAC Name 1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 171°C
Color White
Quantity 1 g
Formula Weight 431.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(tetramethylene)uronium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator