missing translation for 'onlineSavingsMsg'
Learn More

o-Anisoyl chloride, 97%

Catalog No. p-60966
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276110050 5 g Glass bottle
AC276110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276110050 Supplier Thermo Scientific Chemicals Supplier No. 276110050

CAS: 21615-34-9 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21615-34-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00000664
InChI Key RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonym o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
PubChem CID 88969
IUPAC Name 2-methoxybenzoyl chloride
SMILES COC1=CC=CC=C1C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1460g/mL
Boiling Point 128°C to 129°C (8.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5714 to 1.5734
Linear Formula CH3OC6H4COCl
Quantity 5 g
Beilstein 10, 140
Specific Gravity 1.146
Solubility Information Solubility in water: may decompose in water
Formula Weight 170.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material o-Anisoyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-477-5
TSCA TSCA

RUO – Research Use Only