missing translation for 'onlineSavingsMsg'
Learn More

O-Allylhydroxylamine Hydrochloride 98.0+%, TCI America™

$150.98 - $512.62

Chemical Identifiers

CAS 38945-21-0
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD00012957
InChI Key XIQUJVRFXPBMHS-UHFFFAOYSA-N
PubChem CID 3084724
IUPAC Name O-prop-2-enylhydroxylamine;hydrochloride
SMILES C=CCON.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A14491G
View Documents
TCI America
A14491G
1 g N/A
Only null left
 
A14495G
View Documents
TCI America
A14495G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

38945-21-0
109.553
XIQUJVRFXPBMHS-UHFFFAOYSA-N
O-prop-2-enylhydroxylamine;hydrochloride
C3H8ClNO
MFCD00012957
3084724
C=CCON.Cl

Specifications

38945-21-0
White-Yellow
MFCD00012957
XIQUJVRFXPBMHS-UHFFFAOYSA-N
O-prop-2-enylhydroxylamine;hydrochloride
3084724
≥98.0% (T)
O-Allylhydroxylamine Hydrochloride
170°C
C3H8ClNO
1 g
C=CCON.Cl
109.553
109.55
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'