missing translation for 'onlineSavingsMsg'
Learn More

O-Allylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. A14495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14495G 5 g
A14491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14495G Supplier TCI America Supplier No. A14495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38945-21-0
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD00012957
InChI Key XIQUJVRFXPBMHS-UHFFFAOYSA-N
PubChem CID 3084724
IUPAC Name O-prop-2-enylhydroxylamine;hydrochloride
SMILES C=CCON.Cl

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 109.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Allylhydroxylamine Hydrochloride