missing translation for 'onlineSavingsMsg'
Learn More

(+)-O-Acetyl-L-mandelic Acid 98.0+%, TCI America™

Catalog No. A14545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14545G 5 g
A145425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14545G Supplier TCI America Supplier No. A14545G

Chemical Identifiers

CAS 7322-88-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00064215,MFCD00662355,MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym s-2-acetoxy-2-phenylacetic acid, +-o-acetyl-l-mandelic acid, s-+-alpha-acetoxyphenylacetic acid, s-acetylmandelic acid, 2s-2-acetyloxy-2-phenylacetic acid, s-o-acetylmandelic acid, mandelic acid, acetate, l, unii-0n6kt18k98, l-+-o-acetylmandelic acid, s-+-o-acetyl-l-mandelic acid
PubChem CID 6923379
IUPAC Name 2-(acetyloxy)-2-phenylacetic acid
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-O-Acetyl-L-mandelic Acid