missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-O-Acetyl-L-malic Anhydride 97.0+%, TCI America™
$91.33 - $425.00
Chemical Identifiers
| CAS | 59025-03-5 |
|---|---|
| Molecular Formula | C6H6O5 |
| Molecular Weight (g/mol) | 158.109 |
| MDL Number | MFCD00082708 |
| InChI Key | SSWJHSASZZAIAU-BYPYZUCNSA-N |
| Synonym | s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate |
| PubChem CID | 11126528 |
| IUPAC Name | [(3S)-2,5-dioxooxolan-3-yl] acetate |
| SMILES | CC(=O)OC1CC(=O)OC1=O |
Chemical Identifiers
| 59025-03-5 | |
| 158.109 | |
| SSWJHSASZZAIAU-BYPYZUCNSA-N | |
| 11126528 | |
| CC(=O)OC1CC(=O)OC1=O |
| C6H6O5 | |
| MFCD00082708 | |
| s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate | |
| [(3S)-2,5-dioxooxolan-3-yl] acetate |
Specifications
| 59025-03-5 | |
| White-Yellow | |
| MFCD00082708 | |
| s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate | |
| CC(=O)OC1CC(=O)OC1=O | |
| 158.109 | |
| 158.11 | |
| Crystalline Powder |
| 55°C | |
| C6H6O5 | |
| 5 g | |
| SSWJHSASZZAIAU-BYPYZUCNSA-N | |
| [(3S)-2,5-dioxooxolan-3-yl] acetate | |
| 11126528 | |
| ≥97.0% (T) | |
| (-)-O-Acetyl-L-malic Anhydride |
Safety and Handling
TSCA : No