missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Acetyl-L-carnitine Hydrochloride

Catalog No. AAJ6153606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6153606 5 g
AAJ6153614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6153606 Supplier Thermo Scientific Chemicals Supplier No. J6153606

Weak cholinergic agonist | CAS: 5080-50-2 | C9H18ClNO4, C9H17NO4·HCl | 239.70 g/mol

O-Acetyl-L-carnitine hydrochloride is used as a cholinergic agonist that stimulates neuronal response to serotonin and acetylcholine. Acetylcarnitine has antinociceptive activity that may be mediated by enhanced activity of muscarinic cholinergic receptors or mGlu2 glutamate receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5080-50-2
Molecular Formula C9H18ClNO4
Synonym (R)-3-Acetoxy-4-(trimethylammonio)butyrate hydrochloride; L-Carnitine acetyl ester hydrochloride

Specifications

Quantity 5 g
Description White
Content And Storage 2°C to 8°C
For Use With (Application) Weak cholinergic agonist
Formula Weight 239.70g/mol
Sensitivity Hygroscopic
Product Type Signal Transduction Reagent
Form White crystalline powder
Beilstein 4340103
InChI Key JATPLOXBFFRHDN-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC(=O)OC(CC([O-])=O)C[N+](C)(C)C
IUPAC Name hydrogen 3-(acetyloxy)-4-(trimethylazaniumyl)butanoate chloride
Molecular Weight (g/mol) 239.70
Show More Show Less
Recommended Storage Keep cold

RUO – Research Use Only