missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-O-Acetyl-D-mandelic Acid 98.0+%, TCI America™
$74.56 - $229.29
Chemical Identifiers
| CAS | 51019-43-3 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.19 |
| MDL Number | MFCD00004249 |
| InChI Key | OBCUSTCTKLTMBX-UHFFFAOYNA-N |
| Synonym | r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid |
| PubChem CID | 2733814 |
| IUPAC Name | 2-(acetyloxy)-2-phenylacetic acid |
| SMILES | CC(=O)OC(C(O)=O)C1=CC=CC=C1 |
Chemical Identifiers
| 51019-43-3 | |
| 194.19 | |
| OBCUSTCTKLTMBX-UHFFFAOYNA-N | |
| 2733814 | |
| CC(=O)OC(C(O)=O)C1=CC=CC=C1 |
| C10H10O4 | |
| MFCD00004249 | |
| r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid | |
| 2-(acetyloxy)-2-phenylacetic acid |
Specifications
| 51019-43-3 | |
| White | |
| MFCD00004249 | |
| r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid | |
| CC(=O)OC(C(O)=O)C1=CC=CC=C1 | |
| 194.19 | |
| 194.19 | |
| Crystalline Powder |
| 98°C | |
| C10H10O4 | |
| 5 g | |
| OBCUSTCTKLTMBX-UHFFFAOYNA-N | |
| 2-(acetyloxy)-2-phenylacetic acid | |
| 2733814 | |
| ≥98.0% (GC,T) | |
| (-)-O-Acetyl-D-mandelic Acid |
Safety and Handling
TSCA : No