missing translation for 'onlineSavingsMsg'
Learn More

(-)-O-Acetyl-D-mandelic Acid 98.0+%, TCI America™

Catalog No. A14535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14535G 5 g
A145325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14535G Supplier TCI America Supplier No. A14535G

Chemical Identifiers

CAS 51019-43-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid
PubChem CID 2733814
IUPAC Name 2-(acetyloxy)-2-phenylacetic acid
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-O-Acetyl-D-mandelic Acid