missing translation for 'onlineSavingsMsg'
Learn More

o-Acetoacetanisidide 98.0+%, TCI America™

Catalog No. A130625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A130625G 25 g
A1306500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A130625G Supplier TCI America Supplier No. A130625G

Chemical Identifiers

CAS 92-15-9
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD00008781
InChI Key KYYRTDXOHQYZPO-UHFFFAOYSA-N
Synonym n-2-methoxyphenyl-3-oxobutanamide, o-acetoacetanisidide, acetoacet-o-anisidide, n-acetoacetyl-o-anisidine, acetoacetyl-o-anisidide, acetoacetyl-o-anisidine, o-acetoacetaniside, o-methoxyacetoacetanilide, 2-acetoacetylaminoanisole, 2-methoxyacetoacetanilide
PubChem CID 7078
IUPAC Name N-(2-methoxyphenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1OC

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 25 g
Formula Weight 207.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material o-Acetoacetanisidide
EINECSNumber (3)-0689&(3)-1642
RTECSNumber BZ5600000
TSCA Yes