missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7125205
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19265G 5 g
C192625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19265G Supplier TCI America Supplier No. C19265G
Only null left

Chemical Identifiers

CAS 330641-16-2
Molecular Formula C11H15BClF4N5O
Molecular Weight (g/mol) 355.53
MDL Number MFCD04973270
InChI Key GBGVQFJZGHBZMC-UHFFFAOYSA-N
Synonym tctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-6-chloro-1h-benzotriazol-1-yl uronium tetrafluoroborate, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborat e, 6-chlorobenzotriazol-1-yl oxy-dimethylamino methylene-dimethyl-ammonium tetrafluoroborate, tctu; 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium tetrafluoroborate, 6-chloro-1,2,3-benzotriazol-1-yl oxy dimethylamino methylidene dimethylazanium tetrafluoroborate, 1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1, acmc-1agto, ksc569q6h
PubChem CID 24898642
IUPAC Name tetrafluoroboranuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C

Specifications

Melting Point 204°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 355.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(6-Chlorobenzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium Tetrafluoroborate