missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. C19881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19881G 1 g
C19885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19881G Supplier TCI America Supplier No. C19881G

Chemical Identifiers

CAS 330645-87-9
Molecular Formula C11H15ClF6N5OP
Molecular Weight (g/mol) 413.689
InChI Key ZHHGTMQHUWDEJF-UHFFFAOYSA-N
Synonym hctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium hexafluorophosphate, 2-6-chloro-1h-benzotriazole-1-yl-1,1,3,3-tetramethylaminium hexafluorophosphate, o-1h-6-chlorobenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-6-chloro-1-hydrocibenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, pubchem12740, acmc-20aj2u
PubChem CID 42624899
IUPAC Name [(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1.F[P-](F)(F)(F)(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 413.69
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(6-Chlorobenzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium Hexafluorophosphate