missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%

Catalog No. p-7028980
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2640209 10 g
AAH2640218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2640209 Supplier Thermo Scientific Chemicals Supplier No. H2640209

CAS: 330645-87-9 | C11H15ClF6N5OP | 413.689 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330645-87-9
Molecular Formula C11H15ClF6N5OP
Molecular Weight (g/mol) 413.689
MDL Number MFCD04973268
InChI Key ZHHGTMQHUWDEJF-UHFFFAOYSA-N
Synonym hctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium hexafluorophosphate, 2-6-chloro-1h-benzotriazole-1-yl-1,1,3,3-tetramethylaminium hexafluorophosphate, o-1h-6-chlorobenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-6-chloro-1-hydrocibenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, pubchem12740, acmc-20aj2u
PubChem CID 42624899
IUPAC Name [(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼180°C (decomposition)
Quantity 10 g
Formula Weight 413.69
Percent Purity ≥98%
Chemical Name or Material O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only