missing translation for 'onlineSavingsMsg'
Learn More

O-4-Nitrobenzylhydroxylamine Hydrochloride 99.0+%, TCI America™

Catalog No. A55321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55321G 1 g
A55325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55321G Supplier TCI America Supplier No. A55321G

Chemical Identifiers

CAS 2086-26-2
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
MDL Number MFCD00012954
InChI Key LKCAFSOYOMFQSL-UHFFFAOYSA-N
Synonym o-4-nitrobenzyl hydroxylamine hydrochloride, 4-nitrobenzyloxyamine, o-4-nitrophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-4-nitrophenyl methyl-, monohydrochloride, p-nitrobenzyloxyamine hydrochloride, hydroxylamine, o-p-nitrobenzyl-, monohydrochloride, o-p-nitrobenzyl hydroxylamine hydrochloride, o-4-nitrobenzyl hydroxylammonium hydrochloride, 1-aminooxy methyl-4-nitrobenzene hydrochloride, hydroxylamine, o-p-nitrophenylmethyl-, monohydrochloride
PubChem CID 74967
IUPAC Name hydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.61
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-4-Nitrobenzylhydroxylamine Hydrochloride [for HPLC Labeling]
RTECSNumber NC4400050
TSCA Yes