missing translation for 'onlineSavingsMsg'
Learn More

O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%

Catalog No. p-7044323
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0586103 1 g
AAL0586106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0586103 Supplier Thermo Scientific Chemicals Supplier No. L0586103
Only null left

CAS: 2086-26-2 | C7H9ClN2O3 | 204.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2086-26-2
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
MDL Number MFCD00012954
InChI Key LKCAFSOYOMFQSL-UHFFFAOYSA-N
Synonym o-4-nitrobenzyl hydroxylamine hydrochloride, 4-nitrobenzyloxyamine, o-4-nitrophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-4-nitrophenyl methyl-, monohydrochloride, p-nitrobenzyloxyamine hydrochloride, hydroxylamine, o-p-nitrobenzyl-, monohydrochloride, o-p-nitrobenzyl hydroxylamine hydrochloride, o-4-nitrobenzyl hydroxylammonium hydrochloride, 1-aminooxy methyl-4-nitrobenzene hydrochloride, hydroxylamine, o-p-nitrophenylmethyl-, monohydrochloride
PubChem CID 74967
IUPAC Name O-[(4-nitrophenyl)methyl]hydroxylamine;hydrochloride
SMILES [H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∽215°C (decomposition)
Odor Nearly Odorless
Quantity 1 g
Beilstein 3709565
Sensitivity Light and moisture sensitive
Formula Weight 204.61
Percent Purity 98%
Chemical Name or Material O-(4-Nitrobenzyl)hydroxylamine hydrochloride
EINECSNumber 218-228-6
RTECSNumber NC4400050
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only