Learn More
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99%
CAS: 125700-67-6 | C11H16BF4N5O | 321.09 g/mol
Supplier: Thermo Scientific Chemicals L1347006
Description
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.
Solubility
Soluble in acetonitrile (100 mg/ml, clear, colorless), DMF (160.55 mg/ml, clear), and water (3 mg/ml at 20°C).
Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Sensitive to light and moisture. Store under inert gas.
Specifications
| O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate | |
| 195°C to 198°C (decomposition) | |
| MFCD00077413 | |
| 7066325 | |
| Soluble in acetonitrile (100mg/ml,clear,colorless),DMF (160.55mg/ml,clear),and water (3mg/ml at 20°C). | |
| F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C | |
| 321.09 | |
| 321.08 |
| 125700-67-6 | |
| C11H16BF4N5O | |
| 5 g | |
| tbtu, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 2-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-1h-benzotriazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-benzotriazol-1-yl uronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetra methyluronium tetrafluoroborate | |
| JKEKMBGUVUKMQB-UHFFFAOYSA-N | |
| [(1H-1,2,3-benzotriazol-1-yloxy)(dimethylamino)methylidene]dimethylazanium; tetrafluoroboranuide | |
| 2733207 | |
| 99% |
Chemical Identifiers
| 125700-67-6 | |
| 321.09 | |
| JKEKMBGUVUKMQB-UHFFFAOYSA-N | |
| 2733207 |
| C11H16BF4N5O | |
| MFCD00077413 | |
| tbtu, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 2-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-1h-benzotriazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-benzotriazol-1-yl uronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetra methyluronium tetrafluoroborate | |
| F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 423-040-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only