missing translation for 'onlineSavingsMsg'
Learn More

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99%

Catalog No. AAL1347003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1347003 1 g
AAL1347006 5 g
AAL1347014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1347003 Supplier Thermo Scientific Chemicals Supplier No. L1347003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 125700-67-6 | C11H16BF4N5O | 321.09 g/mol

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

Solubility
Soluble in acetonitrile (100 mg/ml, clear, colorless), DMF (160.55 mg/ml, clear), and water (3 mg/ml at 20°C).

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Sensitive to light and moisture. Store under inert gas.

Chemical Identifiers

CAS 125700-67-6
Molecular Formula C11H16BF4N5O
Molecular Weight (g/mol) 321.09
MDL Number MFCD00077413
InChI Key JKEKMBGUVUKMQB-UHFFFAOYSA-N
Synonym tbtu, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 2-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-1h-benzotriazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-benzotriazol-1-yl uronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetra methyluronium tetrafluoroborate
PubChem CID 2733207
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C

Specifications

Melting Point 195°C to 198°C (decomposition)
Quantity 1 g
Beilstein 7066325
Solubility Information Soluble in acetonitrile (100mg/ml,clear,colorless),DMF (160.55mg/ml,clear),and water (3mg/ml at 20°C).
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
Formula Weight 321.08
Percent Purity 99%
Chemical Name or Material O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 423-040-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only