missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%
Description
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.
Solubility
Soluble in acetonitrile.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.
Solubility
Soluble in acetonitrile.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼200°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 7328329 |
| Solubility Information | Soluble in acetonitrile. |
| Formula Weight | 379.25 |
| Percent Purity | 98% |
| Chemical Name or Material | O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 423-020-5 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only