missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, Fisher BioReagents

Catalog No. BP29495
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29495 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29495 Supplier Fisher BioReagents Supplier No. BP2949 5
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Alters membrane permeability and allows potassium ion leakage

Chemical Identifiers

CAS 1400-61-9
Molecular Formula C47H75NO17
Molecular Weight (g/mol) 926.107
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
PubChem CID 133640190
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O

Specifications

Type Nystatin
Physical Form Solid
Source Streptomyces noursei
Loss on Drying < 5.0%
Packaging Amber Glass
Potency > 4400 U/mg
Quantity 5 g
Merck Index 15, 6825
Formula Weight 926.1
Chemical Name or Material Nystatin
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:1