missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Nylon Ties

Catalog No. 19804651
Change view
Click to view available options
Length (Metric):
12.7 cm
For Use With (Application):
For securing tags
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) For Use With (Application)
19804651 12.7 cm For securing tags
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804651 Supplier Accuform Signs Supplier No. MAPT01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

can be used to secure tags or fasten items

Beaded Nylon Ties can be used to secure tags or fasten items.

Specifications

Material Nylon
Color White
Length (Metric) 12.7 cm
Length (English) 5 in.
For Use With (Application) For securing tags