missing translation for 'onlineSavingsMsg'
Learn More

Nylon rod, 3.18mm (0.125 in.) dia, Thermo Scientific™

Catalog No. AA45157BV
Change view
Click to view available options
Quantity:
30 cm
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA45157BV 30 cm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45157BV Supplier Thermo Scientific Chemicals Supplier No. 45157BV

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Nylon rod
Packaging Tube
Quantity 30 cm
MDL Number MFCD00133998