missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Nylon Cap
With PTFE protected silicone liner
Specifications
| Color | Red |
|---|---|
| For Use With (Application) | Organic Synthesis |
| Material | Nylon |
| Quantity | 1 Ea. |
| Product Type | Threaded Screw Cap |