missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Nylon Biopsy Bags
Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.
Supplier: Epredia™ 6774013
Description
- Reduce risk of small specimen loss
- Reduce reagent carryover and pressure artifact often found with sponges
- Remove specimen rapidly with convenient peel-open bag
Dimensions:
- Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
- Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
- Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
Chemical Identifiers
| Biopsy Bag | |
| Nylon | |
| Blue |
Specifications
| Biopsy Bag | |
| Blue | |
| 65 mm | |
| 2.6 in. | |
| 500/Pk. |
| Nylon | |
| 45 mm | |
| 1.8 in. | |
| Medium |